Tisk…

Verapamil hydrochloride ≥99%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 5-((3,4-Dimethoxyphenethyl)(methyl)amino)-2-(3,4-dimethoxyphenyl)-2-isopropylpentanenitrile hydrochloride, Verapamil HCl, 5-[(3,4-Dimethoxyphenethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile Hydrochloride, (±)-Verapamil hydrochloride

ALX550306G001 ALX550306G005
ENZOALX550306G001EA 1570 CZK
ENZOALX550306G001 ENZOALX550306G005
Verapamil hydrochloride ≥99%
Verapamil hydrochloride

Blocks slow voltage-sensitive L-type Ca²⁺ channels. Adrenergic antagonist. Inhibits platelet-activating factor and PMA-stimulated prostaglandin production in Kupffer cells. Induces apoptosis. In vitro resistance modifier in drug resistant human tumor cell lines.


  • Ca2⁺ channel inhibitor


White crystalline powder.


Soluble in 100% ethanol (26mg/ml), methanol or water (83mg/ml).

Vzorec: C₂₇H₃₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00055208
CAS číslo: 152-11-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR