Tisk…
(R)-Roscovitine ≥95% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Roscovitine, (R)-2-((6-(Benzylamino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol
BMLCC2050002 BMLCC2050005
ENZOBMLCC2050002EA 2070 CZK
ENZOBMLCC2050002 ENZOBMLCC2050005
(R)-Roscovitine ≥95% (by TLC)
(R)-Roscovitine
CDK inhibitor
Potent and selective inhibitor of CDKs. Selective for CDK1/cyclin B kinase (IC50=450nM) [2], CDK2 (IC50=700nM) [4] and CDK5/p35 (IC50=160nM) [4,5]. More potent than olomoucine (Prod. No. BML-CC200). Inhibits M phase promoting factor (MPF) kinase activity. Has effects on calcium channel gating.
Vzorec: C₁₉H₂₆N₆O
MW: 354,45 g/mol
MDL Number: MFCD02266401
CAS číslo: 186692-46-6