HC-030031 ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
BMLCA2410010
BMLCA2410050
ENZOBMLCA2410010EA
3140
CZK
ENZOBMLCA2410010
ENZOBMLCA2410050
HC-030031 ≥98% (by HPLC)
HC-030031
A potent and selective TRPA1 inhibitor.
- TRPA1 inhibitor
White solid.
Soluble in DMSO (25 mg/ml).
Vzorec:
C₁₈H₂₁N₅O₃ Storage Temperature: Ambient |
CAS číslo:
349085-38-7 |