Tisk…

AMG-9810 ≥98%

Dodavatel: ENZO LIFE SCIENCES
Varování

BMLCA2320010 BMLCA2320050
ENZOBMLCA2320010EA 3140 CZK
ENZOBMLCA2320010 ENZOBMLCA2320050
AMG-9810 ≥98%
AMG-9810

TRPV1 antagonist.


AMG-9810 is an antagonist of capsaicin activation of TRPV1 (IC₅₀ = 24.5 nM) as well as activation by acid, heat, and endogenous ligands.


Soluble in DMSO (>25 mg/ml), ethanol (20 mg/ml).

Vzorec: C₂₁H₂₃NO₃
MDL Number: MFCD00955103
CAS číslo: 545395-94-6

Objednat

Výsledky specifikačního testu

Purity ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR