Tisk…

Amlodipine ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate

BMLCA2020100 BMLCA2021000
ENZOBMLCA2020100EA 2420 CZK
ENZOBMLCA2020100 ENZOBMLCA2021000
Amlodipine ≥98%
Amlodipine

Ca²⁺ channel blocker.


Amlodipine is a third generation L-type calcium channel blocker with a unique spectrum of actions, over and above its antihypertensive activity which include stimulation of NO synthase, inhibition of monocyte-endothelial adhesion, inhibition of oxidative stress and sustained duration of action.


Soluble in methanol, DMSO (25 mg/ml), 100% ethanol (25 mg/ml), dimethyl formamide (25 mg/ml) or chloroform.

Vzorec: C₂₀H₂₅ClN₂O₅
MW: 408,879 g/mol
MDL Number: MFCD00864687
CAS číslo: 88150-42-9
EINECS: 425-820-1

Objednat

Výsledky specifikačního testu

Purity ≥98% (Assay)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR