Tisk…

Gambogic acid ≥95% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: (Z)-4-((1S,3aR,5S,11R,14aS)-8-Hydroxy-2,2,11-trimethyl-13-(3-methylbut-2-en-1-yl)-11-(4-methylpent-3-en-1-yl)-4,7-dioxo-1,2,5,7-tetrahydro-11H-1,5-methanofuro[3,2-g]pyrano[3,2-b]xanthen-3a(4H)-yl)-2-methylbut-2-enoic acid

BMLAP3050005 BMLAP3050025
ENZOBMLAP3050005EA 3140 CZK
ENZOBMLAP3050005 ENZOBMLAP3050025
Gambogic acid ≥95% (by TLC)
Gambogic acid

Gambogic Acid is a natural product isolated from Garcinia Hurburyi. It induces apoptosis in several tumor cell lines including T47D breast cancer cells and MGC-803 human gastric carcinoma cells but not in normal cells. Apoptosis induction proceeds via upregulation of bax and inhibition of bcl-2 expression.The target of gambogic acid has been identified as the transferrin receptor.


  • Bcl-2 inhibitor, Bax activator


Yellow-orange solid


Soluble in DMSO (25 mg/ml) or ethanol (25 mg/ml)

Vzorec: C₃₈H₄₄O₈
MW: 628,762 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16878985
CAS číslo: 2752-65-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR