Tisk…
Alamethicin ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: U-22324, Antibiotic U-22324
BMLA1500005 BMLA1500025
ENZOBMLA1500005EA 6530 CZK
ENZOBMLA1500005 ENZOBMLA1500025
Alamethicin ≥98% (by TLC)
Alamethicin
Mixture of alamethicin homologs. Membrane permeabilizing antibiotic. Applications include increasing [32P] incorporation into phosphatidylinositol 4-phosphate and permeabilization of sarcoplasmic reticulum vesicles. Causes fusion of lipid vesicles.
  • White to off-white solid
Soluble in DMSO, 100% ethanol or methanol
Balení: Shipping temperature: Ambient
Vzorec: C₉₂H₁₅₀N₂₂O₂₅
MW: 1964,40 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151517
CAS číslo: 59588-86-2