Tisk…
T-2 Toxin ≥98% (by HPLC, TLC)
Dodavatel: ENZO LIFE SCIENCES
ALX630101M001 ALX630101M005
ENZOALX630101M001EA 1570 CZK
ENZOALX630101M001 ENZOALX630101M005
T-2 Toxin ≥98% (by HPLC, TLC)
T-2 Toxin
Monoamine oxidase inhibitor
Mycotoxin. Induces DNA damage and apoptosis. Increases blood-brain barrier permeability and inhibits monoamine oxidase activity in the brain.
Isolated from Fusarium tricinctum.
CAS číslo: 21259-20-1