(Z)-4-Hydroxytamoxifen ≥98%
Dodavatel: ENZO LIFE SCIENCES
ALX550361M001
ALX550361M005
ENZOALX550361M001EA
2360
CZK
ENZOALX550361M001
ENZOALX550361M005
(Z)-4-Hydroxytamoxifen ≥98%
(Z)-4-Hydroxytamoxifen
Tamoxifen metabolite
Metabolite of tamoxifen (Prod. No. ALX-550-095). Exhibits a more potent estrogen agonist/antagonist activity than the parent drug. Has antioxidant properties. Intramembranous inhibitor of lipid peroxidation.
Vzorec:
C₂₆H₂₉NO₂ |
MDL Number:
MFCD00278780 CAS číslo: 68047-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...