Tisk…
6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt, disodium salt ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: CNQX disodium salt, CNQX 2Na
ALX550283M005 ALX550283M010 ALX550283M025
ENZOALX550283M005EA 3050 CZK
ENZOALX550283M005 ENZOALX550283M010 ENZOALX550283M025
6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt, disodium salt ≥98% (by TLC)
6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt
AMPA/kainate receptor antagonist
  • Rust red solid.
Water soluble, potent competitive kainate/quisqualate (non-NMDA) receptor antagonist. More soluble form of CNQX (Prod. No. ALX-550-042).
Soluble in water (5 mg/ml).
Vzorec: C₉H₂N₄O₄.2Na
MW: 276,12 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09953908
CAS číslo: 115066-14-3
Objednat
Výsledky specifikačního testu
Purity (TLC) ≥98%