R-(+)-DIOA ≥98%
Dodavatel: ENZO LIFE SCIENCES
ALX550263M005
ALX550263M025
ENZOALX550263M005EA
6130
CZK
ENZOALX550263M005
ENZOALX550263M025
R-(+)-DIOA ≥98%
R-(+)-DIOA
K⁺ channel blocker
Inhibitor of the K+/Cl--cotransport that does not affect the bumetanide-sensitive Na+/K+/Cl--cotransport system.
CAS číslo:
81166-47-4 |