Tisk…
R-(+)-DIOA ≥98%
Dodavatel: ENZO LIFE SCIENCES
ALX550263M005 ALX550263M025
ENZOALX550263M005EA 6130 CZK
ENZOALX550263M005 ENZOALX550263M025
R-(+)-DIOA ≥98%
R-(+)-DIOA
K⁺ channel blocker
Inhibitor of the K+/Cl--cotransport that does not affect the bumetanide-sensitive Na+/K+/Cl--cotransport system.
CAS číslo: 81166-47-4