Diazoxide ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
|
Varování
|
Synonyms:
3-Methyl-7-chloro-1,2,4-benzothiadiazine 1,1-dioxide, 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-Dioxide
ALX550260G001
ALX550260M500
ENZOALX550260G001EA
9580
CZK
ENZOALX550260G001
ENZOALX550260M500
Diazoxide ≥98% (by HPLC)
Diazoxide
ATP-sensitive K⁺ channel activator.
Effective concentrations in patch clamp experiments of 100 to 600 µM. Blocks desensitisation of AMPA receptors. Acts in pancreatic β-cells, where it is more potent than pinacidil. Antihypertensive and vasodilatory agent.
Soluble in DMSO (25 mg/ml); insoluble in water.
Vzorec:
C₈H₇ClN₂O₂S MW: 230,666 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00078578 CAS číslo: 364-98-7 EINECS: 206-668-1 Merck Index: 14,03004 |
Výsledky specifikačního testu
Purity (HPLC) | ≥98% |
Appearance | White to off-white powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...