SNC80 ≥98% (by HPLC), white to off-white solid
Dodavatel: ENZO LIFE SCIENCES
ALX550172M005
ALX550172M025
ENZOALX550172M005EA
3540
CZK
ENZOALX550172M005
ENZOALX550172M025
SNC80 ≥98% (by HPLC), white to off-white solid
SNC80
delta-opioid receptor agonist
Highly selective and potent non-peptide δ-opioid receptor agonist.
Soluble in 100% ethanol, methanol, DMSO or dichloromethane.
Vzorec:
C₂₈H₃₉N₃O₂ MW: 449.6 g/mol Storage Temperature: Refrigerator |
CAS číslo:
156727-74-1 |
Výsledky specifikačního testu
Purity (HPLC) | ≥98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...