Tisk…

Nifedipine ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester

ALX550091G005 ALX550091G025
ENZOALX550091G005EA 2640 CZK
ENZOALX550091G005 ENZOALX550091G025
Nifedipine ≥98%
Nifedipine

Ca²⁺ channel inhibitor


Voltage-sensitive (L-type) Ca²⁺ channel blocker. Vasodilator. Induces apoptosis.

Vzorec: C₁₇H₁₈N₂O₆
MW: 346.34 g/mol
MDL Number: MFCD00057326
CAS číslo: 21829-25-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR