Tisk…
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt
ALX480003G001 ALX480003M050 ALX480003M250
ENZOALX480003G001EA 8270 CZK
ENZOALX480003G001 ENZOALX480003M050 ENZOALX480003M250
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Vzorec: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37
Storage Temperature: Freezer
MDL Number: MFCD00065390
CAS číslo: 24292-60-2
EINECS: 246-129-8