Tisk…
Nonactin (from Streptomyces griseus) ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: Polynactin, Werramycin A
ALX450008M005 ALX450008M025
ENZOALX450008M005EA 1540 CZK
ENZOALX450008M005 ENZOALX450008M025
Nonactin (from Streptomyces griseus) ≥98% (by TLC)
Nonactin
Macrotetrolide antibiotic. Monovalent cation ionophore with high selectivity for ammonium and potassium. Specifically inhibits the processing of cytoplasmic precursor proteins destined for the mitochondria. Inhibits the effects on P170 glycoprotein-mediated efflux of chemotherapeutic agents in multidrug resistant (MDR) cancer cells.
  • Monovalent cation ionophore
White to off-white crystalline solid.
Soluble in DMSO (10mg/ml), dichloromethane (10mg/ml), methanol, 100% ethanol; insoluble in water.
Vzorec: C₄₀H₆₄O₁₂
MW: 736,9 g/mol
Storage Temperature: Freezer
CAS číslo: 6833-84-7