Tisk…

Ionomycin calcium salt

Dodavatel: ENZO LIFE SCIENCES

ALX450007M001 ALX450007M005
ENZOALX450007M001EA 4710 CZK
ENZOALX450007M001 ENZOALX450007M005
Ionomycin calcium salt
Ionomycin calcium salt
Vzorec: C₄₁H₇₀CaO₉
MDL Number: MFCD00151183
CAS číslo: 56092-82-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR