Necrox-5™ methanesulphonate
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
[5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine
Necrosis inhibitor
Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models.
Vzorec:
C₂₅H₃₁N₃O₃S·2CH₄O₃S MW: 645.8 g/mol |
CAS číslo:
1383718-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...