Tisk…
Zoledronic acid ≥98% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: (1-Hydroxy-2-(1H-imidazol-1-yl)ethane-1,1-diyl)diphosphonic acid, ZOL-446, CGP 4244, CGP 42446A, Zoledronate
ALX430153G001 ALX430153M025 ALX430153M100
ENZOALX430153G001EA 28390 CZK
ENZOALX430153G001 ENZOALX430153M025 ENZOALX430153M100
Zoledronic acid ≥98% (by HPLC)
Zoledronic acid
Acid sphingomyelinase inhibitor
Inhibitor of osteoclastic bone resorption. Prevents osteoporosis and skeletal fractures, particularly in patients with multiple myeloma or prostate cancer. Also used to treat hypercalcemia.Inhibition of acid sphingomyelinase (aSMase) with an IC50 5µm.
Synthetic.
Vzorec: C₅H₁₀N₂O₇P₂
MW: 272,09 g/mol
MDL Number: MFCD00867791
CAS číslo: 118072-93-8