MnTBAP chloride ≥97%
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
MnTBAP, [5,10,15,20-Tetrakis(4-carboxyphenyl)-21H,23H-porphine]manganese(III) chloride
ALX430069M010
ALX430069M050
ENZOALX430069M010EA
1450
CZK
ENZOALX430069M010
ENZOALX430069M050
MnTBAP chloride ≥97%
MnTBAP chloride
SOD mimetic. Peroxynitrite scavenger.
Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Inhibits neuronal apoptosis.
Vzorec:
C₄₈H₂₈MnN₄O₈Cl MW: 879.2 g/mol |
MDL Number:
MFCD06227760 CAS číslo: 55266-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...