Tisk…

R59 949 ≥98%

Dodavatel: ENZO LIFE SCIENCES

ALX430028M005 ALX430028M025
ENZOALX430028M005EA 5510 CZK
ENZOALX430028M005 ENZOALX430028M025
R59 949 ≥98%
R59 949

Potentiates PKC activity


Inhibitor of diacylglycerol kinase in intact cells (IC50=0.12µM) and in isolated membranes (IC50=0.3µM). More potent analog of R59 022 (Prod. No. ALX-430-027). Useful for elucidating roles of protein kinase C (PKC).

Vzorec: C₂₈H₂₅F₂N₃OS
MDL Number: MFCD00069258
CAS číslo: 120166-69-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR