NOR-3 ≥97% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Nitric oxide donor
Non-thiol-based nitric oxide (NO) donor. NOR-3 shows a variety of biological and pharmacological effects. Strong vasodilatory effects in rat aorta, rabbit aorta and dog coronary artery; potent inhibitory activities against antiplatelet aggregation and thrombus formation; effective aggregation inhibition of human platelet raising by ADP; cardioprotective effects in the ischemia/reperfusion system; antianginal effect.
CAS číslo:
163180-49-2 |