Tisk…

HAT activator, CTPB ≥97% (by 1H-NMR)

Dodavatel: ENZO LIFE SCIENCES

ALX420033M001 ALX420033M005
ENZOALX420033M001EA 2110 CZK
ENZOALX420033M001 ENZOALX420033M005
HAT activator, CTPB ≥97% (by 1H-NMR)
HAT activator, CTPB

p300 HAT activator.


Potent activator of p300 HAT (histone acetyltransferase), but not of PCAF (p300/CBP-associated factor) HAT activities.


Soluble in DMSO.

Vzorec: C₃₁H₄₃ClF₃NO₂
Storage Temperature: Freezer
MDL Number: MFCD08702707
CAS číslo: 586976-24-1

Objednat

Výsledky specifikačního testu

Purity ≥97% (¹H-NMR)
Identity Determined by NMR


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR