Tisk…

BAY 41-2272

Dodavatel: ENZO LIFE SCIENCES

Synonyms: 5-Cyclopropyl-2-(1-(2-fluorobenzyl)-1H-pyrazolo[3,4-b]pyridin-3-yl)pyrimidin-4-amine

ALX420030M005 ALX420030M025
ENZOALX420030M005EA 4710 CZK
ENZOALX420030M005 ENZOALX420030M025
BAY 41-2272
BAY 41-2272

Guanylyl cyclase activator


NO-independent activator of soluble guanylyl cyclase (sGC). Activates both isoforms alpha1beta1 and alpha1beta2 of sGC. While the related compound YC-1 (Prod. No. ALX-420-025) also acts as a non-specific phosphodiesterase inhibitor, BAY 41-2272 has no effect on phosphodiesterases. Stimulation of sGC is not blocked by high concentrations of NO scavengers (e.g. PTIO, Prod. No. ALX-430-007) and the combination of BAY 41-2272 with the NO donor DEA NONOate (Prod. No. ALX-430-034) potentiates the activation of sGC. Although BAY 41-2272 alone is not as strong a stimulator of sGC as NO, concentrations as low as 10-100nM stimulate sGC to a level that would be expected to cause biologically important increases in cGMP. ODQ (Prod. No. ALX-270-034), a potent and selective inhibitor of sGC, completely inhibited the effect of BAY 41-2272. The activity of BAY 41-2272 is described in selected literature references.

Vzorec: C₂₀H₁₇FN₆
MDL Number: MFCD06411391
CAS číslo: 256376-24-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR