Tisk…
Cyclophosphamide monohydrate (synthetic) ≥97% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Nebezpečí
Synonyms: 2-(Bis(2-chloroethyl)amino)-1,3,2-oxazaphosphinane 2-oxide hydrate, Cyclophosphamide hydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-1,3,2-oxazaphosphorin-2-oxide
ALX400051G001 ALX400051G005
ENZOALX400051G001EA 1450 CZK
ENZOALX400051G001 ENZOALX400051G005
Cyclophosphamide monohydrate (synthetic) ≥97% (by HPLC)
Cyclophosphamide monohydrate
Alkylates DNA, forming DNA-DNA cross-links that result in inhibition of DNA synthesis and cell death.
  • DNA alkylating agent
  • White to off-white crystalline solid
  • Soluble in water; slightly soluble in 100% ethanol
Interestingly the repair enzyme O6-alkylguanine-DNA alkyltransferase, long thought to be involved with resistance to methylating and chloroethylating agents, may also contribute to resistance to the cytotoxic and mutagenic effects of cyclophosphamide.
Informace pro objednávání: Shipped at ambient temperature
Vzorec: C₇H₁₇Cl₂N₂O₃P
MW: 279.1 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00149395
CAS číslo: 6055-19-2
UN: 3464
ADR: 6.1 III