Tisk…
Ciprofloxacin ≥98%
Dodavatel: ENZO LIFE SCIENCES
Synonyms: 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic Acid
ALX380287G005 ALX380287G025
ENZOALX380287G005EA 1450 CZK
ENZOALX380287G005 ENZOALX380287G025
Ciprofloxacin ≥98%
Ciprofloxacin
Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.
  • Effective against Mycobacterium tuberculosis
  • Potentially anxiogenic
White to off-white powder
Soluble in dilute hydrochloric acid; insoluble in water
Balení: Shipping temperature: Ambient
Vzorec: C₁₇H₁₈FN₃O₃
MW: 331.35 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00185755
CAS číslo: 85721-33-1