Tisk…

Cefotaxime sodium salt ≥95% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

ALX380271G001 ALX380271M250
ENZOALX380271G001EA 4790 CZK
ENZOALX380271G001 ENZOALX380271M250
Cefotaxime sodium salt ≥95% (by HPLC)
Cefotaxime sodium salt

Inhibits bacterial cell wall synthesis. Effective against Gram-negative and Gram-positive bacteria.


  • Cephalosporin antibiotic
  • Resistant against β-lactamase


White to light yellow powder


Soluble in water (>50 mg/mL)


Balení: Shipping temperature: Ambient

Vzorec: C₁₆H₁₇N₅O₇S₂
Storage Temperature: Freezer
MDL Number: MFCD00079073
CAS číslo: 64485-93-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR