Tisk…

Verruculogen ≥96% (by HPLC, TLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: NA 209A

ALX380056M001 ALX380056M005
ENZOALX380056M001EA 5390 CZK
ENZOALX380056M001 ENZOALX380056M005
Verruculogen ≥96% (by HPLC, TLC)
Verruculogen

Cell cycle inhibitor


Neurotoxin that acts as an inhibitor of the M phase of the mammalian cell cycle. Inhibits Ca²⁺-activated K+ channels. Decreases GABA levels in CNS.


Isolated from Penicillium verruculosum.

Vzorec: C₂₇H₃₃N₃O₇
MW: 511.6 g/mol
CAS číslo: 12771-72-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR