Tisk…

Oligomycin (from Streptomyces diastatochromogenes) ≥90%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: Oligomycin A, B, C, Oligomycin complex

ALX380037M005 ALX380037M010
ENZOALX380037M005EA 3510 CZK
ENZOALX380037M005 ENZOALX380037M010
Oligomycin (from Streptomyces diastatochromogenes) ≥90%
Oligomycin

F-ATPase inhibitor.


Macrolide antibiotic that inhibits membrane bound mitochondrial ATPase (F1), preventing phosphoryl group transfer. Induces apoptosis.


White to faint yellow powder


Soluble in 100% ethanol, methanol (10 mg/mL) or acetone (50 mg/mL)


Balení: Shipping temperature: Ambient

Vzorec: C₄₅H₇₄O₁₁
MW: 789,3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01779388
CAS číslo: 1404-19-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR