Tisk…
Pseudohypericin ≥98% (by HPLC, TLC)
Dodavatel: ENZO LIFE SCIENCES
ALX350276M001 ALX350276M005
ENZOALX350276M001EA 5900 CZK
ENZOALX350276M001 ENZOALX350276M005
Pseudohypericin ≥98% (by HPLC, TLC)
Pseudohypericin
Inhibitor of PKC and dopamine beta-hydroxylase
Inhibitor of protein kinase C (PKC) (IC50=15µg/ml). Antiviral and antiretroviral activity. Potent inhibitor of dopamine-beta-hydroxylase (IC50=3µg/ml).
Isolated from Hypericum perforatum.
CAS číslo: 55954-61-5