Orlistat ≥97%
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
N-Formyl-L-leucine (1S)-1-[[(2S,3S)-3-Hexyl-4-oxo-2-oxetanyl]methyl]dodecyl Ester, (S)-(S)-1-((2S,3S)-3-Hexyl-4-oxooxetan-2-yl)tridecan-2-yl 2-formamido-4-methylpentanoate
Fatty acid synthase inhibitor
Cell permeable, irreversible inhibitor of gastric and pancreatic lipases. Shows only minimal activity against amylase, trypsin, chymotrypsin, or phospholipase A2 (PLA2). Partially inhibits the hydrolysis of triglycerides and lowers the absorption of dietary fat and promotes weight loss. Anti-obesity drug. Exhibits antitumor activity by inhibition of the thioesterase domain of fatty acid synthase (FAS) both in vitro and in vivo.
Synthetic. Originally isolated from Streptomyces sp.
Vzorec:
C₂₉H₅₃NO₅ MW: 495,75 g/mol |
MDL Number:
MFCD05662360 CAS číslo: 96829-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...