Tisk…

Nodularin

Dodavatel: ENZO LIFE SCIENCES

ALX350061C100 ALX350061C250 ALX350061M001
ENZOALX350061C100EA 5820 CZK
ENZOALX350061C100 ENZOALX350061C250 ENZOALX350061M001
Nodularin
Nodularin

Inhibitor of PP1 and PP2A


Inhibitor of protein phosphatase 1 (PP1) (IC50=1.8nM), protein phosphatase 2A (PP2A) (IC50=0.026nM) and to a lesser extent protein phosphatase 2B (PP2B) (IC50=8.7µm). Similar to microcystin-LR (Prod. No. ALX-350-012) but with increased water solubility. Genotoxic. Hepatoxic. Induces oxidative DNA damage by oxidation of purines. Induces apoptosis in HepG2 cells.


Isolated from Nodularia spumigena.

Vzorec: C₄₁H₆₀N₈O₁₀
MW: 824,977 g/mol
CAS číslo: 118399-22-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR