Tisk…
L-Canavanine sulphate ≥99% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: Canavanine sulphate, L-α-Amino-γ-(guanidinooxy)-n-butyric acid sulphate
ALX350002M100 ALX350002M500
ENZOALX350002M100EA 1670 CZK
ENZOALX350002M100 ENZOALX350002M500
L-Canavanine sulphate ≥99% (by TLC)
L-Canavanine sulphate
The non-protein amino acid L-canavanine is an analog of L-arginine. Selective inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Induces apoptotic cell death and shows antiproliferative and immunotoxic effects.
  • iNOS inhibitor
White to-off white powder.
Soluble in water (100mg/ml).
Vzorec: C₅H₁₂N₄O₃·H₂SO₄
MW: 274.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012618
CAS číslo: 2219-31-0