Tisk…

Imatinib mesylate ≥98% (by HPLC)

Dodavatel: ENZO LIFE SCIENCES

Synonyms: N-(4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)phenyl)-4-((4-methylpiperazin-1-yl)methyl)benzamide methanesulphonate, Imatinib Methanesulphonate

ALX270492M025 ALX270492M500
ENZOALX270492M025EA 1890 CZK
ENZOALX270492M025 ENZOALX270492M500
Imatinib mesylate ≥98% (by HPLC)
Imatinib mesylate

Tyrosine kinase inhibitor.


  • White to pale yellow crystalline powder.
  • Soluble in water(200 mg/ml); DMSO(100 mg/ml); DMF(10 mg/ml); PBS(2 mg/ml) and sparingly soluble in ethanol(0,2 mg/ml).


Highly specific for BCR-ABL, the enzyme associated with chronic myelogenous leukemia (CML) and certain forms of acute lymphoblastic leukemia (ALL). Also shown to inhibit the transmembrane receptor kit and platelet-derived growth factor (PDGF) receptors.

Vzorec: C₂₉H₃₁N₇O·CH₄SO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04307699
CAS číslo: 220127-57-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR