Tisk…

AS-252424 ≥98% (by 1H-NMR)

Dodavatel: ENZO LIFE SCIENCES

ALX270465M001 ALX270465M005
ENZOALX270465M001EA 1760 CZK
ENZOALX270465M001 ENZOALX270465M005
AS-252424 ≥98% (by 1H-NMR)
AS-252424

Potent and selective phosphoinositide 3-kinase (PI(3)K) p110γ inhibitor shown to be selective for class IB PI(3)K-mediated cellular effects.

Vzorec: C₁₄H₈FNO₄S
MDL Number: MFCD11045281
CAS číslo: 900515-16-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR