RO-3306 ≥95% (by 1H-NMR)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
(Z)-5-(Quinolin-6-ylmethylene)-2-((thiophen-2-ylmethyl)amino)thiazol-4(5H)-one, (Z)-5-(Quinolin-6-ylmethylene)-2-[(thiophen-2-ylmethyl)amino]thiazol-4(5H)-one
CDK/cyclin inhibitor
Inhibitor of CDK1/cyclin B1 and CDK1/cyclin A. Reversibly arrests human cells at the G2/M border of the cell cycle. Allows for effective cell synchronisation in early mitosis. Downregulates expression of the antiapoptotic proteins Bcl-2 and survivin and blocks p53-mediated induction of p21 and MDM2.
Vzorec:
C₁₈H₁₃N₃OS₂ |
MDL Number:
MFCD17392573 CAS číslo: 872573-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...