Tisk…
PI103 ≥97% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: PI-103, 3-(4-Morpholinopyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol
ALX270460M001 ALX270460M005
ENZOALX270460M001EA 2040 CZK
ENZOALX270460M001 ENZOALX270460M005
PI103 ≥97% (by HPLC)
PI103
Potent, cell permeable and ATP-competitive inhibitor of DNA-PK, phosphoinositide 3-kinase (PI(3)K), the rapamycin-sensitive (mTORC1) and rapamycin-insensitive (mTORC2) complexes of the protein kinase mTOR. Exhibits little activity towards a panel of more than 40 other kinases even at concentrations as high as 10µM. Effectively blocks cell proliferation in glioma cell lines both in vitro and in vivo.
  • Inhibitor of DNA-PK, PI3 kinase and mTOR
Off-white solid.
Soluble in DMSO (5mg/ml; warm).
Vzorec: C₁₉H₁₆N₄O₃
Storage Temperature: Freezer
MDL Number: MFCD11983145
CAS číslo: 371935-74-9