RepSox ≥97% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
ALX270445M001
ALX270445M005
ENZOALX270445M001EA
3400
CZK
ENZOALX270445M001
ENZOALX270445M005
RepSox ≥97% (by HPLC)
RepSox
Cell permeable, potent, selective and ATP-competitive inhibitor of TGF-β RI kinase (IC50=23nM, 4nM and 18nM for binding, auto-phosphorylation and cellular assay in HepG2 cells of TGF-β RI kinase, respectively). Minimally affects a panel of 9 closely related kinases including p38 MAPK (IC50>16µM).
White to off-white or yellow solid.
5mg/ml soluble in DMSO or methanol.
Vzorec:
C₁₇H₁₃N₅ Storage Temperature: Freezer |
MDL Number:
MFCD09037561 CAS číslo: 446859-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...