LY 303511 ≥95% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
8-phenyl-2-piperazin-1-ylchromen-4-one, 8-phenyl-2-(piperazin-1-yl)-4H-chromen-4-one, EM 101, NV 128, 2-(1-Piperazinyl)-8-phenyl-4H-1-benzopyran-4-one
ALX270410M001
ALX270410M005
ENZOALX270410M001EA
3310
CZK
ENZOALX270410M001
ENZOALX270410M005
LY 303511 ≥95% (by HPLC)
LY 303511
Useful negative control for the PI(3)K (phosphoinositide 3-kinase) inhibitor LY-294,002. Does not inhibit PI(3)K even at concentrations.
Yellow solid.
Soluble in DMSO.
Vzorec:
C₁₉H₁₈N₂O₂ MW: 306,36 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03453556 CAS číslo: 154447-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...