Anacardic acid ≥95% (by 1H-NMR)
Dodavatel: ENZO LIFE SCIENCES
Cell permeable salicylic acid analog that acts as a potent, non-competitive inhibitor of p300 and PCAF (p300/CBP-associated factor) histone acetyltransferase (HAT) activities (IC50~8.5µM and ~5µM, respectively). Displays a variety of other biological activities, such as antibacterial, antimicrobial, prostaglandin synthase inhibition, tyrosinase, and lipoxygenase inhibition.Inhibits SUMOylation of RanGAP1-C2 with an IC50 = 2.2µM
Synthetic.
Vzorec:
C₂₂H₃₆O₃ |
MDL Number:
MFCD07368254 CAS číslo: 16611-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...