Tisk…

MS-275 ≥95% (by 1H-NMR)

Dodavatel: ENZO LIFE SCIENCES

ALX270378M001 ALX270378M025
ENZOALX270378M001EA 2480 CZK
ENZOALX270378M001 ENZOALX270378M025
MS-275 ≥95% (by 1H-NMR)
MS-275

HDAC1 inhibitor


Preferentially inhibits HDAC1 (IC50=300nM) over HDAC3 (IC50=8µM). Has no inhibitory activity towards HDAC8 (IC50>100µM).

Vzorec: C₂₁H₂₀N₄O₃
MDL Number: MFCD08272435
CAS číslo: 209783-80-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR