Tisk…

SH-5

Dodavatel: ENZO LIFE SCIENCES

ALX270349M001 ALX270349MC05
ENZOALX270349M001EA 12940 CZK
ENZOALX270349M001 ENZOALX270349MC05
SH-5
SH-5

Akt inhibitor


Inhibitor of Akt (PKB) activation without affecting activation of the upstream kinase PDK-1, or other kinases downstream of Ras such as MAPK. Induces apoptosis and kills a variety of cancer cell lines containing high levels of active Akt and depending upon Akt for survival.

Vzorec: C₂₉H₅₉O₁₀P
MDL Number: MFCD08703608
CAS číslo: 701976-54-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR