Tisk…

BEC ammonium salt ≥95% (by TLC)

Dodavatel: ENZO LIFE SCIENCES

ALX270345M001 ALX270345M005
ENZOALX270345M001EA 1950 CZK
ENZOALX270345M001 ENZOALX270345M005
BEC ammonium salt ≥95% (by TLC)
BEC ammonium salt

Slow-binding competitive inhibitor of arginase I (Ki=0.4-0.6µM & KD=2.22µM for recombinant rat liver arginase) and arginase II (Ki=0.31µM at pH 7.5 and 0.03µM at pH 9.5 for human recombinant type II arginase). Enhances NO-dependent smooth muscle relaxation in human penile corpus cavernosum tissue. Does not inhibit nitric oxide synthase (NOS) and may serve as a valuable reagent to probe the physiological relationship between arginase and NOS. Inhibitor of rat aortic smooth muscle cell proliferation.


  • Arginase inhibitor


White to off-white powder.


Soluble in water (10mg/ml).

Vzorec: C₅H₁₂BNO₄S·NH₄
MW: 192.0 · 18.0 g/mol
Storage Temperature: Refrigerator
CAS číslo: 63107-40-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR