Aminopurvalanol A ≥97% (by 1H-NMR)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
(R)-2-((6-((3-Amino-5-chlorophenyl)amino)-9-isopropyl-9H-purin-2-yl)amino)-3-methylbutan-1-ol
ALX270249M001
ALX270249M005
ENZOALX270249M001EA
2420
CZK
ENZOALX270249M001
ENZOALX270249M005
Aminopurvalanol A ≥97% (by 1H-NMR)
Aminopurvalanol A
CDK/cyclin inhibitor
Cell permeable, reversible and competitive inhibitor of CDK1/cyclin B (IC50=33 nM), CDK2/cyclin A (IC50=33 nM), CDK2/cyclin E (IC50=28 nM) and CDK5/p35 (IC50=20 nM).
Vzorec:
C₁₉H₂₆ClN₇O MW: 403.9090 g/mol |
MDL Number:
MFCD06798384 CAS číslo: 220792-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...