Tisk…

Purvalanol A ≥99% (by NMR)

Dodavatel: ENZO LIFE SCIENCES

ALX270246M001 ALX270246M005
ENZOALX270246M001EA 2800 CZK
ENZOALX270246M001 ENZOALX270246M005
Purvalanol A ≥99% (by NMR)
Purvalanol A

Inhibitor of human CDK1 (IC50=4nM), CDK2/cyclin A (IC50=70nM), Cdc2/cyclin B, CDK2/cyclin E (IC50=35nM), CDK4/cyclin D1 (IC50=850nM) as well as CDK5/p35 (IC50=75nM). More membrane permeable than purvalanol B.


  • CDK/cyclin inhibitor


Off-white to yellow powder.


Soluble in DMSO, methanol or dichloromethane.

Vzorec: C₁₉H₂₅N₆OCl
Storage Temperature: Freezer
MDL Number: MFCD02179211
CAS číslo: 212844-53-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR