Tisk…
Caffeic acid phenylethyl ester ≥97% (by HPLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms: (E)-Phenethyl 3-(3,4-dihydroxyphenyl)acrylate
ALX270244M010 ALX270244M050
ENZOALX270244M010EA 1420 CZK
ENZOALX270244M010 ENZOALX270244M050
Caffeic acid phenylethyl ester ≥97% (by HPLC)
Caffeic acid phenylethyl ester
HIV-1 integrase inhibitor
Active component of propolis from honeybee hives, known to have anti-mitogenic, anti-carcinogenic, anti-inflammatory, and immunomodulatory properties. Potent and specific inhibitor of NF-kappaB activation, induced by TNF-alpha (Prod. No. ALX-522-008 (human) or Prod. No. ALX-522-009 (mouse)), PMA (Prod. No. BML-PE160) and other inflammatory agents. Exhibits inhibitory activity against HIV-1 integrase (IC50=7µM). Suppresses lipid peroxidation and inhibits ornithine decarboxylase, protein tyrosine kinase and lipoxygenase activities. Induces apoptosis.
Synthetic.
Vzorec: C₁₇H₁₆O₄
MW: 284,31 g/mol
MDL Number: MFCD00866470
CAS číslo: 115610-29-2