Vinyl-L-NIO ≥95% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
Synonyms:
L-VNIO, N5-(1-Imino-3-butenyl)-L-ornithine
ALX270216M025
ENZOALX270216M025EA
14540
CZK
ENZOALX270216M025
Vinyl-L-NIO ≥95% (by TLC)
Vinyl-L-NIO
Very selective and potent neuronal nitric oxide synthase (nNOS/NOS I) inhibitor.
- nNOS inhibitor
Clear film in the bottom of the vial.
Soluble in water, 100% ethanol (30mg/ml), DMSO (50mg/ml) or dimethyl formamide (50mg/ml).
Vzorec:
C₉H₁₇N₃O₂·HCl MW: 235,7 g/mol Storage Temperature: Refrigerator |
CAS číslo:
728944-69-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...