Tisk…

Ebselen

Dodavatel: ENZO LIFE SCIENCES
Nebezpečí

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

ALX270097M025
ENZOALX270097M025EA 3420 CZK
ENZOALX270097M025
Ebselen
Ebselen
Vzorec: C₁₃H₉NOSe
MW: 274,20 g/mol
MDL Number: MFCD00210937
CAS číslo: 60940-34-3
UN: 3283
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR