Tisk…
Cantharidin ≥98% (by TLC)
Dodavatel: ENZO LIFE SCIENCES
ALX270063M025 ALX270063M100
ENZOALX270063M025EA 2940 CZK
ENZOALX270063M025 ENZOALX270063M100
Cantharidin ≥98% (by TLC)
Cantharidin
PP2A inhibitor
Potent protein phosphatase 2A (PP2A) inhibitor. At higher concentrations, also inhibits protein phosphatase 1 (PP1).
Vzorec: C₁₀H₁₂O₄
MW: 196.202 g/mol
MDL Number: MFCD00134968
CAS číslo: 56-25-7