Tisk…

Bisindoylmaleimide X hydrochloride ≥98%

Dodavatel: ENZO LIFE SCIENCES

Synonyms: Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride

ALX270057M001 ALX270057M005
ENZOALX270057M001EA 3510 CZK
ENZOALX270057M001 ENZOALX270057M005
Bisindoylmaleimide X hydrochloride ≥98%
Bisindoylmaleimide X hydrochloride

PKC inhibitor


Selective inhibitor of protein kinase C (PKC).

Vzorec: C₂₆H₂₄N₄O₂·HCl
MW: 461 g/mol
MDL Number: MFCD04972010
CAS číslo: 145317-11-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR